Fmoc-2-amino-4,5-dimethoxybenzoic acid

Supplier: Matrix Scientific

041709-1G 041709-5G
101852-864EA 378.46 USD
101852-864 101852-866
Fmoc-2-amino-4,5-dimethoxybenzoic acid
Fmoc-2-amino-4,5-dimethoxybenzoic acid

Hazard: Irritant

Formula: C₂₄H₂₁NO₆
MDL Number: MFCD02682187
CAS Number: 855005-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR