Fmoc-2-amino-3-methylbenzoic acid

Supplier: Matrix Scientific

041708-1G 041708-5G
101852-860EA 498.46 USD
101852-860 101852-862
Fmoc-2-amino-3-methylbenzoic acid
Fmoc-2-amino-3-methylbenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₄
MW: 373.41 g/mol
MDL Number: MFCD02682220
CAS Number: 885951-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR