Fmoc-2-amino-3-methoxybenzoic acid

Supplier: Matrix Scientific

041707-1G 041707-5G
101852-856EA 686.15 USD
101852-856 101852-858
Fmoc-2-amino-3-methoxybenzoic acid
Fmoc-2-amino-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₅
MDL Number: MFCD02682213
CAS Number: 372144-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR