Fmoc-2-amino-3,5-dimethylbenzoic acid

Supplier: Matrix Scientific

041706-1G 041706-5G
101852-852EA 686.15 USD
101852-852 101852-854
Fmoc-2-amino-3,5-dimethylbenzoic acid
Fmoc-2-amino-3,5-dimethylbenzoic acid

Hazard: Irritant

Formula: C₂₄H₂₁NO₄
MDL Number: MFCD02682185
CAS Number: 1185302-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR