Fmoc-2-amino-3,4-dimethylbenzoic acid
Supplier: Matrix Scientific
041705-1G
041705-5G
101852-848EA
498.46
USD
101852-848
101852-850
Fmoc-2-amino-3,4-dimethylbenzoic acid
Fmoc-2-amino-3,4-dimethylbenzoic acid
Hazard: Irritant
Formula:
C₂₄H₂₁NO₄ |
MDL Number:
MFCD09750476 CAS Number: 1185302-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...