Di-fmoc-3,5-diaminobenzoic acid

Supplier: Matrix Scientific

041703-1G 041703-5G
101852-840EA 1115.38 USD
101852-840 101852-842
Di-fmoc-3,5-diaminobenzoic acid
Di-fmoc-3,5-diaminobenzoic acid

Hazard: Irritant

Formula: C₃₇H₂₈N₂O₆
MDL Number: MFCD02682247
CAS Number: 248602-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR