Di-fmoc-3,4-diaminobenzoic acid

Supplier: Matrix Scientific

Synonyms: 3,4-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)benzoic acid

041702-1G 041702-5G
101852-836EA 249.23 USD
101852-836 101852-838
Di-fmoc-3,4-diaminobenzoic acid
Di-fmoc-3,4-diaminobenzoic acid

Hazard: Irritant

Formula: C₃₇H₂₈N₂O₆
MW: 596.65 g/mol
MDL Number: MFCD02682246
CAS Number: 345958-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR