Di-boc-3,5-diaminobenzoic acid
Supplier: Matrix Scientific
Synonyms:
3,5-Bis((tert-butoxycarbonyl)amino)benzoic acid
041701-1G
041701-5G
101852-832EA
164.62
USD
101852-832
101852-834
Di-boc-3,5-diaminobenzoic acid
Di-boc-3,5-diaminobenzoic acid
Hazard: Irritant
Formula:
C₁₇H₂₄N₂O₆ MW: 352.39 g/mol |
MDL Number:
MFCD00270356 CAS Number: 133887-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...