Di-boc-3,4-diaminobenzoic acid

Supplier: Matrix Scientific

Synonyms: 3,4-Bis((tert-butoxycarbonyl)amino)benzoic acid

041700-1G 041700-5G
101852-828EA 364.62 USD
101852-828 101852-830
Di-boc-3,4-diaminobenzoic acid
Di-boc-3,4-diaminobenzoic acid

Hazard: Irritant

Formula: C₁₇H₂₄N₂O₆
MW: 352.39 g/mol
MDL Number: MFCD02682245
CAS Number: 66636-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR