Boc-5-amino-2-methoxybenzoic acid
Supplier: Matrix Scientific
041699-1G
041699-5G
101852-824EA
275.38
USD
101852-824
101852-826
Boc-5-amino-2-methoxybenzoic acid
Boc-5-amino-2-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₃H₁₇NO₅ |
MDL Number:
MFCD04972601 CAS Number: 1075242-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...