Boc-5-amino-2-chlorobenzoic acid

Supplier: Matrix Scientific

041698-1G 041698-5G
101852-820EA 326.15 USD
101852-820 101852-822
Boc-5-amino-2-chlorobenzoic acid
Boc-5-amino-2-chlorobenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₄
MDL Number: MFCD02682166
CAS Number: 503555-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR