Boc-5-acetamido-2-aminobenzoic acid

Supplier: Matrix Scientific

041697-1G 041697-5G
101852-816EA 326.15 USD
101852-816 101852-818
Boc-5-acetamido-2-aminobenzoic acid
Boc-5-acetamido-2-aminobenzoic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₅
MDL Number: MFCD02682143
CAS Number: 848126-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR