Boc-4-methylaminobenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid

041696-1G 041696-5G
101852-812EA 144.62 USD
101852-812 101852-814
Boc-4-methylaminobenzoic acid ≥95%
Boc-4-methylaminobenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
MDL Number: MFCD04972273
CAS Number: 263021-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR