4-((tert-Butoxycarbonyl)amino)benzoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
Boc-4-Abz-OH, 4-(Boc-amino)benzoic Acid
041694-5G
041694-25G
101852-804EA
32.31
USD
101852-804
101852-806
4-((tert-Butoxycarbonyl)amino)benzoic acid ≥95%
4-((tert-Butoxycarbonyl)amino)benzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₅NO₄ MW: 237.3 g/mol |
MDL Number:
MFCD00037428 CAS Number: 66493-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...