Boc-4-amino-3-methylbenzoic acid

Supplier: Matrix Scientific

Synonyms: 4-((tert-Butoxycarbonyl)amino)-3-methylbenzoic acid

041693-1G 041693-5G
101852-800EA 250.77 USD
101852-800 101852-802
Boc-4-amino-3-methylbenzoic acid
Boc-4-amino-3-methylbenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄
MDL Number: MFCD02682225
CAS Number: 180976-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR