Boc-4-amino-3-methoxybenzoic acid

Supplier: Matrix Scientific

041692-1G 041692-5G
101852-796EA 423.08 USD
101852-796 101852-798
Boc-4-amino-3-methoxybenzoic acid
Boc-4-amino-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₅
MDL Number: MFCD02682217
CAS Number: 180976-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR