Boc-4-amino-2-chlorobenzoic acid

Supplier: Matrix Scientific

Synonyms: 4-((tert-Butoxycarbonyl)amino)-2-chlorobenzoic acid

041691-1G 041691-5G
101852-792EA 515.38 USD
101852-792 101852-794
Boc-4-amino-2-chlorobenzoic acid
Boc-4-amino-2-chlorobenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₄
MW: 271.7 g/mol
MDL Number: MFCD02682164
CAS Number: 232275-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR