3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid

Supplier: Matrix Scientific

Synonyms: 3-(BOC-Aminomethyl)benzoic acid, 3-[[(tert-butoxycarbonyl)amino]methyl]benzoic acid, N-Boc-3-Aminomethyl-benzoicacid

041690-1G 041690-5G
101852-788EA 224.62 USD
101852-788 101852-790
3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid
3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄
MW: 251.282 g/mol
MDL Number: MFCD00273426
CAS Number: 117445-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR