Boc-3-Abz-OH

Supplier: Matrix Scientific

Synonyms: 3-{[(tert-Butoxy)carbonyl]amino}benzoic acid, Boc-3-aminobenzoic acid

041689-1G 041689-5G
101852-784EA 15.38 USD
101852-784 101852-786
Boc-3-Abz-OH
Boc-3-Abz-OH

Hazard: Irritant

Formula: C₁₂H₁₅NO₄
MW: 237.26 g/mol
MDL Number: MFCD00235884
CAS Number: 111331-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR