Boc-3-aminobenzene-1,2-dicarboxylic acid

Supplier: Matrix Scientific

041688-1G 041688-5G
101852-780EA 326.15 USD
101852-780 101852-782
Boc-3-aminobenzene-1,2-dicarboxylic acid
Boc-3-aminobenzene-1,2-dicarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₆
MDL Number: MFCD02682147
CAS Number: 1185302-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR