Boc-3-amino-4-chlorobenzoic acid
Supplier: Matrix Scientific
Synonyms:
3-((tert-Butoxycarbonyl)amino)-4-chlorobenzoic acid
041685-1G
041685-5G
101852-768EA
326.15
USD
101852-768
101852-770
Boc-3-amino-4-chlorobenzoic acid
Boc-3-amino-4-chlorobenzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₄ClNO₄ MW: 271.7 g/mol |
MDL Number:
MFCD02682162 CAS Number: 160450-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...