Boc-3-amino-2-methylbenzoic acid

Supplier: Matrix Scientific

041684-1G 041684-5G
101852-764EA 326.15 USD
101852-764 101852-766
Boc-3-amino-2-methylbenzoic acid
Boc-3-amino-2-methylbenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
MDL Number: MFCD01861303
CAS Number: 143617-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR