Boc-2-amino-6-fluorobenzoic acid

Supplier: Matrix Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-6-fluorobenzoic acid

041679-1G 041679-5G
101852-748EA 536.92 USD
101852-748 101852-750
Boc-2-amino-6-fluorobenzoic acid
Boc-2-amino-6-fluorobenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄FNO₄
MDL Number: MFCD02682192
CAS Number: 1048919-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR