Boc-2-amino-6-chlorobenzoic acid

Supplier: Matrix Scientific

041678-1G 041678-5G
101852-744EA 326.15 USD
101852-744 101852-746
Boc-2-amino-6-chlorobenzoic acid
Boc-2-amino-6-chlorobenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₄
MDL Number: MFCD02682160
CAS Number: 616224-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR