Boc-2-amino-5-methoxybenzoic acid

Supplier: Matrix Scientific

041676-1G 041676-5G
101852-736EA 686.15 USD
101852-736 101852-738
Boc-2-amino-5-methoxybenzoic acid
Boc-2-amino-5-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₅
MDL Number: MFCD02682214
CAS Number: 262614-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR