Boc-2-amino-5-iodobenzoic acid

Supplier: Matrix Scientific

041675-1G 041675-5G
101852-732EA 446.15 USD
101852-732 101852-734
Boc-2-amino-5-iodobenzoic acid
Boc-2-amino-5-iodobenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄INO₄
MW: 363.15 g/mol
MDL Number: MFCD02682210
CAS Number: 445479-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR