Boc-2-amino-5-fluorobenzoic acid

Supplier: Matrix Scientific

041674-1G 041674-5G
101852-728EA 536.92 USD
101852-728 101852-730
Boc-2-amino-5-fluorobenzoic acid
Boc-2-amino-5-fluorobenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄FNO₄
MDL Number: MFCD02682190
CAS Number: 141940-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR