Boc-2-amino-5-chlorobenzoic acid

Supplier: Matrix Scientific

Synonyms: 2-tert-Butoxycarbonylamino-5-chlorobenzoic acid, 2-((tert-Butoxycarbonyl)amino)-5-chlorobenzoic acid, 2-[(tert-butoxycarbonyl)amino]-5-chlorobenzoic acid

041673-1G 041673-5G
101852-724EA 326.15 USD
101852-724 101852-726
Boc-2-amino-5-chlorobenzoic acid
Boc-2-amino-5-chlorobenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₄
MW: 271.7 g/mol
MDL Number: MFCD02682158
CAS Number: 253677-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR