5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid
Supplier: Matrix Scientific
041672-1G
041672-5G
101852-720EA
326.15
USD
101852-720
101852-722
5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid
5-Bromo-2-((tert-butoxycarbonyl)amino)benzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₄BrNO₄ |
MDL Number:
MFCD01619006 CAS Number: 306937-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...