Boc-2-amino-3-methylbenzoic acid

Supplier: Matrix Scientific

Synonyms: 2-tert-Butoxycarbonylamino-3-methylbenzoic acid

041668-1G 041668-5G
101852-704EA 498.46 USD
101852-704 101852-706
Boc-2-amino-3-methylbenzoic acid
Boc-2-amino-3-methylbenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄
MDL Number: MFCD02682219
CAS Number: 669713-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR