Boc-2-amino-3-methoxybenzoic acid

Supplier: Matrix Scientific

041667-1G 041667-5G
101852-700EA 686.15 USD
101852-700 101852-702
Boc-2-amino-3-methoxybenzoic acid
Boc-2-amino-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₅
MDL Number: MFCD02682212
CAS Number: 1026003-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR