Boc-2-amino-3,5-dimethylbenzoic acid
Supplier: Matrix Scientific
Synonyms:
2-{[(tert-Butoxy)carbonyl]amino}-3,5-dimethylbenzoic acid, 2-tert-Butoxycarbonylamino-3,5-dimethylbenzoic acid
041666-1G
041666-5G
101852-696EA
686.15
USD
101852-696
101852-698
Boc-2-amino-3,5-dimethylbenzoic acid
Boc-2-amino-3,5-dimethylbenzoic acid
Hazard: Irritant
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol |
MDL Number:
MFCD02682184 CAS Number: 669713-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...