Boc-2-amino-3,4-dimethylbenzoic acid

Supplier: Matrix Scientific

041665-1G 041665-5G
101852-692EA 498.46 USD
101852-692 101852-694
Boc-2-amino-3,4-dimethylbenzoic acid
Boc-2-amino-3,4-dimethylbenzoic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD09750475
CAS Number: 1185295-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR