Boc-2-amino-3,4,5-trimethoxybenzoic acid

Supplier: Matrix Scientific

041664-1G 041664-5G
101852-688EA 686.15 USD
101852-688 101852-690
Boc-2-amino-3,4,5-trimethoxybenzoic acid
Boc-2-amino-3,4,5-trimethoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₇
MDL Number: MFCD02682243
CAS Number: 1185297-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR