3,5-Bis[2-(fmoc-amino)ethoxy]benzoic acid

Supplier: Matrix Scientific

041661-1G 041661-5G
101852-676EA 686.15 USD
101852-676 101852-678
3,5-Bis[2-(fmoc-amino)ethoxy]benzoic acid
3,5-Bis[2-(fmoc-amino)ethoxy]benzoic acid

Hazard: Irritant

Formula: C₄₁H₃₆N₂O₈
MDL Number: MFCD09750477
CAS Number: 1185295-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR