3,5-Bis[2-(boc-amino)ethoxy]benzoic acid

Supplier: Matrix Scientific

Synonyms: 3,5-Bis(2-((tert-butoxycarbonyl)amino)ethoxy)benzoic acid

041660-1G 041660-5G
101852-672EA 686.15 USD
101852-672 101852-674
3,5-Bis[2-(boc-amino)ethoxy]benzoic acid
3,5-Bis[2-(boc-amino)ethoxy]benzoic acid

Hazard: Irritant

Formula: C₂₁H₃₂N₂O₈
MW: 440.5 g/mol
MDL Number: MFCD04974222
CAS Number: 187960-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR