Fmoc-threo-β-methyl-DL-aspartic acid

Supplier: Matrix Scientific

041658-250MG 041658-1G
101852-664EA 275.38 USD
101852-664 101852-666
Fmoc-threo-β-methyl-DL-aspartic acid
Fmoc-threo-beta-methyl-DL-aspartic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₆
MW: 369.38 g/mol
MDL Number: MFCD02682506
CAS Number: 1219167-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR