Fmoc-iminodiacetic acid

Supplier: Matrix Scientific

041656-1G 041656-5G
101852-656EA 164.62 USD
101852-656 101852-658
Fmoc-iminodiacetic acid
Fmoc-iminodiacetic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₆
MDL Number: MFCD01321017
CAS Number: 112918-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR