Fmoc-γ-methylene-DL-glutamic acid

Supplier: Matrix Scientific

041655-100MG 041655-250MG
101852-652EA 958.46 USD
101852-652 101852-654
Fmoc-γ-methylene-DL-glutamic acid
Fmoc-gamma-methylene-DL-glutamic acid

Hazard: Irritant

Formula: C₂₁H₁₉NO₆
MW: 381.39 g/mol
MDL Number: MFCD02682500
CAS Number: 1219174-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR