Fmoc-L-Gla(OtBu)2-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-γ-carboxy-Glu(OtBu)₂-OH, Fmoc-Gla(OtBu)₂-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-4-(tert-butoxycarbonyl)-5-oxopentanoic acid

041654-250MG 041654-1G
101852-648EA 823.08 USD
101852-648 101852-650
Fmoc-L-Gla(OtBu)2-OH
Fmoc-L-Gla(OtBu)2-OH

Hazard: Irritant

Formula: C₂₉H₃₅NO₈
MDL Number: MFCD00038770
CAS Number: 111662-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR