Fmoc-DL-2-aminoheptanedioic acid

Supplier: Matrix Scientific

041651-1G 041651-5G
101852-634EA 1192.31 USD
101852-634 101852-636
Fmoc-DL-2-aminoheptanedioic acid
Fmoc-DL-2-aminoheptanedioic acid

Hazard: Irritant

Formula: C₂₂H₂₃NO₆
MW: 397.43 g/mol
MDL Number: MFCD09750510
CAS Number: 161125-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR