Fmoc-D-2-aminoadipic acid
Supplier: Matrix Scientific
041649-1G
041649-5G
101852-626EA
498.46
USD
101852-626
101852-628
Fmoc-D-2-aminoadipic acid
Fmoc-D-2-aminoadipic acid
Hazard: Irritant
Formula:
C₂₁H₂₁NO₆ |
MDL Number:
MFCD01317727 CAS Number: 218457-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...