Fmoc-L-α-aminosuberic acid
Supplier: Matrix Scientific
041648-1G
041648-5G
101852-622EA
661.54
USD
101852-622
101852-624
Fmoc-L-α-aminosuberic acid
Fmoc-L-alpha-aminosuberic acid
Hazard: Irritant
Formula:
C₂₃H₂₅NO₆ MW: 411.46 g/mol |
MDL Number:
MFCD01317728 CAS Number: 218457-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...