Fmoc-α-methyl-L-aspartic acid
Supplier: Matrix Scientific
041647-100MG
041647-500MG
101852-618EA
536.92
USD
101852-618
101852-620
Fmoc-α-methyl-L-aspartic acid
Fmoc-alpha-methyl-L-aspartic acid
Hazard: Irritant
Formula:
C₂₀H₁₉NO₆ MW: 369.38 g/mol |
MDL Number:
MFCD02682502 CAS Number: 1217830-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...