Fmoc-α-methyl-D-aspartic acid

Supplier: Matrix Scientific

041645-100MG 041645-500MG
101852-610EA 536.92 USD
101852-610 101852-612
Fmoc-α-methyl-D-aspartic acid
Fmoc-alpha-methyl-D-aspartic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₆
MW: 369.38 g/mol
MDL Number: MFCD02682504
CAS Number: 1283766-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR