Fmoc-[N-α-(4-aminobenzoyl)]-L-glutamic acid

Supplier: Matrix Scientific

041643-1G 041643-5G
101852-602EA 386.15 USD
101852-602 101852-604
Fmoc-[N-α-(4-aminobenzoyl)]-L-glutamic acid
Fmoc-[N-alpha-(4-aminobenzoyl)]-L-glutamic acid

Hazard: Irritant

Formula: C₂₇H₂₄N₂O₇
MW: 488.5 g/mol
MDL Number: MFCD12547712
CAS Number: 1562435-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR