Fmoc-(N-γ-ethyl)-L-glutamine

Supplier: Matrix Scientific

041642-1G 041642-5G
101852-598EA 686.15 USD
101852-598 101852-600
Fmoc-(N-γ-ethyl)-L-glutamine
Fmoc-(N-gamma-ethyl)-L-glutamine

Hazard: Irritant

Formula: C₂₂H₂₄N₂O₅
MW: 396.45 g/mol
MDL Number: MFCD02682670
CAS Number: 676230-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR