Boc-threo-β-methyl-DL-aspartic acid

Supplier: Matrix Scientific

041641-250MG 041641-1G
101852-594EA 275.38 USD
101852-594 101852-596
Boc-threo-β-methyl-DL-aspartic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₆
MDL Number: MFCD02682505
CAS Number: 159107-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR