Boc-L-2-aminoadipic acid

Supplier: Matrix Scientific

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)hexanedioic acid

041640-1G 041640-5G
101852-590EA 498.46 USD
101852-590 101852-592
Boc-L-2-aminoadipic acid
Boc-L-2-aminoadipic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₆
MW: 261.28 g/mol
MDL Number: MFCD00063354
CAS Number: 77302-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR