Boc-γ-methylene-DL-glutamic acid

Supplier: Matrix Scientific

041638-100MG 041638-250MG
101852-582EA 958.46 USD
101852-582 101852-584
Boc-γ-methylene-DL-glutamic acid
Boc-gamma-methylene-DL-glutamic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₆
MDL Number: MFCD02682499
CAS Number: 175088-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR